Order Entry
Belgium
Orders LinkContactUsLinkComponent
Detectable plaster rolls

Detectable plaster rolls

Detectable plaster roll, blue, 1 m, 1000×60 mm
Catalog # SOEH1009090
Supplier:   SOEHNGEN
Detectable plaster rolls
Catalog # SOEH1009090
Supplier:  SOEHNGEN
Detectable plaster rolls

Detectable plaster roll, blue, 1 m, 1000×60 mm

Price
13,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

These detectable plasters are used to cover small skin lesions. The aluminium strip under the wound pad make the plasters detectable by all conventional metal detectors. Ideal for certain areas of production, in particular the food industry.

  • Skin-friendly and reliable acrylic adhesive
  • Water resistant, hypoallergenic and breathable
  • Blue colour ensures easy visual recognition
Specifications
  • Description:
    Detectable plaster roll, blue, 1 m
  • L×W:
    1000×60 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.