Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Superdex 200 Increase small-scale SEC columns

Superdex™ 200 Increase 3.2/300 GL, Glass, 3,2×300 mm, 2,4 ml, 0,004 - 0,050 ml, <0,15 ml/min
  28-9909-46
 :   Cytiva
Superdex 200 Increase small-scale SEC columns
  28-9909-46
 :  Cytiva
Superdex 200 Increase small-scale SEC columns
Superdex 200 Increase small-scale SEC columns

Superdex™ 200 Increase 3.2/300 GL, Glass, 3,2×300 mm, 2,4 ml, 0,004 - 0,050 ml, <0,15 ml/min

 
3 110,00 €
   
1 * 1 ST
 

Superdex 200 Increase size exclusion chromatography columns are well suited for high-resolution analysis and small-scale purification of monoclonal antibodies (mAb) and other biomolecules with Mr~ 10000 to 600000.

  • Enhanced performance: Improved resolution and runtime compared to predecessor Superdex 200
  • Very high resolution: Small bead size and narrow particle size distribution provide high resolution, for high protein purity
  • High flow rates: Rigid beads give excellent pressure/flow properties

Superdex 200 Increase is a next-generation, dextran-agarose composite matrix for SEC. With smaller and more rigid beads than their predecessors, Superdex Increase columns deliver higher resolution purification in shorter run times.

 
  • Flow rate:
    <0,15 ml/min
  • Capacity:
    2,4 ml
  • Description:
    Superdex™ 200 Increase 3.2/300 GL
  • Seq:
    01
  • Column hardware:
    Glass
  • Bed dimensions:
    3,2×300 mm
  • Sample volume:
    0,004 - 0,050 ml
  • Cat. No.:
    28-9909-46
  • Max. pressure:
    30 bar
  • Storage:
    4 to 30 °C, 20% Ethanol
  • pH range:
    3 - 12