Ion selective electrodes, Fluoride, IntelliCAL™
Product Details & Documents
IntelliCAL™ ISEF121 is a digital combination fluoride ion selective electrode (ISE) with a non-refillable, single junction reference and built-in temperature sensor. The solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
- Compatible with digital Hach meters only (e.g. HQd meters)
- Requires virtually no maintenance
- Fast, stable, and accurate results
- Ultimate traceability in measurement history
- Can be moved between meters without the need to re-calibrate or re-enter measurement settings
The probe is available with a 1 or 3 meter cable and is intended for laboratory use. Ideal for measuring fluoride concentrations in drinking water, wastewater, and general water quality applications.
Ordering information: Electrode is supplied with integrated temperature sensor, calibration certificate, and basic user manual.
The analysis package contains the IntelliCAL™ ISEF12101 electrode, fluoride ionic strength adjustor powder pillows (100), fluoride 0,5 mg/L standard solution (500 ml), fluoride 1,0 mg/L standard solution (500 ml), and fluoride 2,0 mg/L standard solution (500 ml).
Specifications
- Concentration:0,01 - 19000 mg/l F-
- Connection:3 m cable
- Ø×L:12×220 mm
- Type:IntelliCAL™ ISEF121 (Fluoride)
- Description:IntelliCAL™ fluoride electrode ISEF121, Ag/AgCl reference system
- Material:Epoxy
- Seq:51
- Diaphr.:Porous PTFE annular ring
- Electrolyte:driTEK gel (non refillable)
- Temp.:5...50 °C
Accessories
Product Information
- Catalog #TypePkAvailabilityPrice
- Fluoride ionic strength adjustor (ISA) powder pillows, 0,01 mg/L to saturatedFluoride ionic strength adjustor (ISA) powder pillows, 0,01 mg/L to saturatedHACH258999PP100Log in for availability1 * 100 ST - 132,00 €
Product Information
- Catalog #ConcentrationAccuracyAvailabilityPrice
- Fluoride standard solutionFluoride standard solutionHACH291491,00 µg/ml±0,02 µg/mlLog in for availability1 * 500 mL - 40,20 €
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



