Orders LinkContactUsLinkComponent
Test kits, digital titration method
Iron (Total) reagent set, high range, 100 - 1000 mg/l, 1
N° de catalogue HACH2449300
Fournisseur:   Hach
Test kits, digital titration method
N° de catalogue HACH2449300
Fournisseur:  Hach
Numéro CAS:  
Test kits, digital titration method
Test kits, digital titration method
Iron (Total) reagent set, high range, 100 - 1000 mg/l, 1
Numéro CAS:  
Prix
236,00 €
Log in for availability
1 * 1 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
 : Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Pk:
    1
  • Description:
    Iron (Total) reagent set, high range
  • Range:
    100 - 1000 mg/l
Specifications