Orders LinkContactUsLinkComponent
Caps, LABOCAP, without grip
15-16 mm, Green
  LUDI184110635
 :   LUDI
Caps, LABOCAP, without grip
  LUDI184110635
 :  LUDI
Caps, LABOCAP, without grip
Caps, LABOCAP, without grip
15-16 mm, Green
17,40 €
1 * 10 ST
Made of anodised aluminum with stainless steel spring clip.
  • Springs clamp against the outer wall of the vessel
  • Anodised and coloured
  • Autoclavable
Without grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
  • For Ø ext.:
    15-16 mm
  • Colour:
    Green