Buffer solution pH 7, Phosphate buffer
Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution, 7,00, ±0,02, Phosphate, Plastic bottle, 100 mL
Supplier: Thermo Fisher Scientific
Buffer solution pH 7, Phosphate buffer
Catalog # ACRO258591000
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Documents
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Phosphate
- Pk:100 mL
- Description:Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution
- pH accuracy:±0,02
- pH value:7,00
- Cat. No.:ACRO258591000
- Concentration:3.54 g potassium dihydrogen phosphate and 14.7 g i-sodium hydrogen phosphate per litre
- Appearance:Clear colourless solution



