Orders LinkContactUsLinkComponent
Trypsin protease, MS grade
Trypsin protease, MS grade
Trypsin Protease, 5×20 µg, 1 KIT
Catalog # PIER90057
Trypsin protease, MS grade
Catalog # PIER90057
CAS Number:  
Trypsin protease, MS grade
Trypsin protease, MS grade
Trypsin Protease, 5×20 µg, 1 KIT
CAS Number:  
Price
111,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Pierce™ Trypsin Protease, MS Grade, is highly purified porcine trypsin that has been chemically modified for maximum activity and stability in proteomic applications.
  • Cleaves at the carboxyl side of lysine and arginine residues with greater than 95% specificity
  • No detectable chymotrypsin activity thus making it highly pure
  • Chemically modified for reduced autolytic activity
Applications include in-gel digestion of proteins from 1-D or 2-D gels and in-solution tryptic digestion of proteins. The selectivity of this enzyme is critical for reproducible protein digestion and mass spectrometry-based protein identification. It has been treated with TPCK to eliminate chymotryptic activity, improving digestion specificity. The enzyme is most active in pH ranges 7 to 9 and can be reversibly inactivated at pH <4.
Documents
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Trypsin Protease, 5×20 µg