ContactUsLinkComponent
Taneo universal light STZL 12 R, 15W/100-240v/50/60Hz/IP20, aluminium anod. (ZO), Screen : MILKY, Colour temp.: 5000 K, Mains plug SEP.: CEE 7/XVI Fastening : scerw-down flange, table clamp (accessory), wall bracket (accessory) 1 * 1 items
Catalog # WLDM112576000/1
Supplier:  WALDMANN
Taneo universal light STZL 12 R, 15W/100-240v/50/60Hz/IP20, aluminium anod. (ZO), Screen : MILKY, Colour temp.: 5000 K, Mains plug SEP.: CEE 7/XVI Fastening : scerw-down flange, table clamp (accessory), wall bracket (accessory) 1 * 1 items
Catalog # WLDM112576000/1
Supplier:  WALDMANN
Frequently Bought Together