ContactUsLinkComponent
tray shelf, chipboard/polyethylene, melamin resin laminated/raw load capacity (evenly distributed load) (kg): 50 volume (l): 33 for medium and tall cabinets with width 1200 mm for model(s): q90, s90 1 * 1 items
Catalog # MSGA30027
Supplier:  asecos
tray shelf, chipboard/polyethylene, melamin resin laminated/raw load capacity (evenly distributed load) (kg): 50 volume (l): 33 for medium and tall cabinets with width 1200 mm for model(s): q90, s90 1 * 1 items
Catalog # MSGA30027
Supplier:  asecos
Frequently Bought Together