Visitors coats, 4400
White, M, 167 - 176 cm, 167-176 cm, 81 - 91 cm
Catalog # MMMA4400WM
Supplier: 3M
Jump to:
Product Details & Documents
These disposable coats made of soft, breathable PP are designed to protect the wearer against non hazardous dusts and non hazardous light liquid splashes.
- Low-linting properties
- Breathable material to help reduce heat build-up
- Front zip closing (single way)
- Elasticated cuffs for a secure fit
- Internal pocket on the wearers left-hand side
Typical applications include: General maintenance, catering, electronics manufacturing, laboratories and hygiene applications in general.
Colour: White
Weight: 45 g/m²
Specifications
- Clothing Size:M
- Colour:White
- Height:167 - 176 cm
- Length:167-176 cm
- Seq:06
- Chest size:81 - 91 cm
- Cat. No.:MMMA4400WM
- Garment application:Laboratory|Food
- Gender:Unisex
- Pockets:With pockets
- Closure type:Zip closure
- Weight:45 g/m²



