Visitors coats, 4400
White, 4XL, 200 - 206 cm, 117 - 132 cm
Catalog # MMMA4400W4XL
Supplier: 3M
Jump to:
Product Details & Documents
These disposable coats made of soft, breathable PP are designed to protect the wearer against non hazardous dusts and non hazardous light liquid splashes.
- Low-linting properties
- Breathable material to help reduce heat build-up
- Front zip closing (single way)
- Elasticated cuffs for a secure fit
- Internal pocket on the wearers left-hand side
Typical applications include: General maintenance, catering, electronics manufacturing, laboratories and hygiene applications in general.
Colour: White
Weight: 45 g/m²
Specifications
- Clothing Size:4XL
- Colour:White
- Height:200 - 206 cm
- Seq:11
- Chest size:117 - 132 cm
- Cat. No.:MMMA4400W4XL
- Garment application:Laboratory|Food
- Gender:Unisex
- Pockets:With pockets
- Closure type:Zip closure
- Weight:45 g/m²



