Visitors coats, 4400
White, 3XL, 194 - 200 cm, 194-200 cm, 109 - 117 cm
Catalog # MMMA4400W3XL
Supplier: 3M
Jump to:
Product Details & Documents
These disposable coats made of soft, breathable PP are designed to protect the wearer against non hazardous dusts and non hazardous light liquid splashes.
- Low-linting properties
- Breathable material to help reduce heat build-up
- Front zip closing (single way)
- Elasticated cuffs for a secure fit
- Internal pocket on the wearers left-hand side
Typical applications include: General maintenance, catering, electronics manufacturing, laboratories and hygiene applications in general.
Colour: White
Weight: 45 g/m²
Specifications
- Clothing Size:3XL
- Colour:White
- Height:194 - 200 cm
- Length:194-200 cm
- Seq:10
- Chest size:109 - 117 cm
- Cat. No.:MMMA4400W3XL
- Garment application:Laboratory|Food
- Gender:Unisex
- Pockets:With pockets
- Closure type:Zip closure
- Weight:45 g/m²
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



