Specimen containers, disposable
Product Details & Documents
These containers and closures are made of PP. They are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples.
- Disposable
- Resist temperatures from −20 to + 80 °C
Tight-fitting lids prevent leaks and odors. Certain models listed below have a tight-fitting lid. Also offered with an incorporate tamperproof lid which is opened by first removing tab on side of lid, then by lifting the cover.
The 2600 and 5700 ml sizes are provided with a plastic handle to facilitate handling.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
SIMPC580-5_TDS_EN
Datasheet
Specifications
- Ø opening:200 mm
- Capacity:2600 ml
- Height:124 mm
- Seq:07
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



