Trolleys, serving
Serving trolley, heavy duty, with reinforced chassis, SSW, 895×595×1649 mm, 33,7 kg, 5
Catalog # HUPF0112262
Supplier: HUPFER METALLWERKE
Jump to:
Product Details & Documents
Transport trolley complete with 25 mm deep shelves. Shelves complete with bevelled edges for user comfort. With four swivel castors, two of them with brake.
- Corrosion resistant
- Disc bumpers are made of elastic and shock resistant plastic material, it protect trolley from damage
- Maintenance free
High-quality stainless steel construction.
Documents
HUPF0111932_TDS_EN
Datasheet
Specifications
- Description:Serving trolley, heavy duty, with reinforced chassis, SSW
- W×D×H:895×595×1649 mm
- Shelves:5
- Weight:33,7 kg
Accessories
Product Information
- Catalog #AvailabilityPrice
- Gallery, stainless steel, four sided, height 50 mm, for 600×400 mm shelvesGallery, stainless steel, four sided, height 50 mm, for 600×400 mm shelvesHUPF0114507Log in for availability1 * 1 ST - 248,00 €
Product Information
- Catalog #AvailabilityPrice
- Gallery, stainless steel, height: 50 mm, width: 799 mm, depth: 430 mm, for 800×500 mm shelvesGallery, stainless steel, height: 50 mm, width: 799 mm, depth: 430 mm, for 800×500 mm shelvesHUPF0114502Log in for availability1 * 1 ST - 228,00 €



