ContactUsLinkComponent
Tube capillary microhaematocrit non-heparinised 75uL, interior 1.1-1.2mm, exterior 1.5-1.6mm, made of high grade quality glass in compliance with DIN ISO 12 772, length: approx. 75mm ± 0.5mm, wall thickness: approx. 0.2 ± 0.025mm 1 * 1.000 items
Catalog # MARI2900000
Supplier:  MARIENFELD
Tube capillary microhaematocrit non-heparinised 75uL, interior 1.1-1.2mm, exterior 1.5-1.6mm, made of high grade quality glass in compliance with DIN ISO 12 772, length: approx. 75mm ± 0.5mm, wall thickness: approx. 0.2 ± 0.025mm 1 * 1.000 items
Catalog # MARI2900000
Supplier:  MARIENFELD
Frequently Bought Together