Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium deoxycholate

Sodium deoxycholate

Glass bottle for solids, 1 kg
Catalog # A1531.1000
Irritant
Supplier:   PanReac AppliChem
Sodium deoxycholate
Catalog # A1531.1000
Supplier:  PanReac AppliChem
CAS Number:  302-95-4
Sodium deoxycholate

Glass bottle for solids, 1 kg

CAS Number:  302-95-4
Price
1 230,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
  • CAS Number:302-95-4
  • Formula:C₂₄H₃₉NaO₄
  • MDL Number:MFCD00064139
  • Molecular Weight:414.56 g/mol
  • Melting Pt:357…365 °C
  • Storage Temperature:Ambient
  • EINECS:206-132-7

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications