VWR®, Tip Boxes, Empty
Tip-Box N, PP, with push on lid, stackable and autoclavable, for 200 µl tips, 2 - 200 µl tips
613-2287
Documents
VWR® for Liquid Handling
Product Literature
- Description:Tip-Box N, PP, with push on lid, stackable and autoclavable, for 200 µl tips
- Seq:01
- For use with:2 - 200 µl tips