Orders LinkContactUsLinkComponent
Accessories
Accessories
Upgrade AD (demineralised water), All Miele glassware units
Catalog # 141-0504
Accessories
Catalog # 141-0504
Supplier:  
Accessories
Accessories
Upgrade AD (demineralised water), All Miele glassware units
Discontinued
Product Details & Documents
  • For lower and upper baskets
  • With 7 x 7 mm perforations
  • Stainless steel
Specifications
  • Description:
    Upgrade AD (demineralised water)
  • Seq:
    48
  • For:
    All Miele glassware units