Order Entry
Belgium
Orders LinkContactUsLinkComponent
Process documentaion

Process documentaion

APH 407 Network cable for connection to the network, length 5 m (type 7), Transparent silver, G 7823 , G 7824, G 7825, G 7826, G 7831, PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593
Catalog # 141-0624
Supplier:   MIELE
Process documentaion
Catalog # 141-0624
Supplier:  MIELE
Process documentaion
Process documentaion

APH 407 Network cable for connection to the network, length 5 m (type 7), Transparent silver, G 7823 , G 7824, G 7825, G 7826, G 7831, PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593

Price
41,04 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    APH 407 Network cable for connection to the network, length 5 m (type 7), Transparent silver
  • Seq:
    19
  • For:
    G 7823 , G 7824, G 7825, G 7826, G 7831, PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593
  • Cat. No.:
    141-0624
  • W×D×H:
    250×350×30 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.