Orders LinkContactUsLinkComponent
Inserts
E 136 suitable for 56 half petri dishes with Ø 100 mm, equipped with 56 holders, height 70 mm, distance approx. 26 mm
Catalog # 141-0380
Supplier:   MIELE
Inserts
Catalog # 141-0380
Supplier:  MIELE
Inserts
Inserts
E 136 suitable for 56 half petri dishes with Ø 100 mm, equipped with 56 holders, height 70 mm, distance approx. 26 mm
Price
436,05 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 136 suitable for 56 half petri dishes with Ø 100 mm, equipped with 56 holders, height 70 mm, distance approx. 26 mm
  • Seq:
    49