Order Entry
Belgium
Orders LinkContactUsLinkComponent
Microscope slides

Microscope slides

Microscope slides with ground edges, 1,0 mm, 76×26 mm
Catalog # HECH42401010
Microscope slides
Catalog # HECH42401010
Microscope slides
Microscope slides

Microscope slides with ground edges, 1,0 mm, 76×26 mm

Price
122,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Pre-cleaned slides with either cut or ground edges.

Specifications
  • Description:
    Microscope slides with ground edges
  • L×W:
    76×26 mm
  • Thickness:
    1,0 mm