Orders LinkContactUsLinkComponent
Microplates, 96-well, non binding
Microplates, 96-well, non binding
96-well microplate, V-bottom, non binding, solid bottom, Clear
Catalog # 738-0166
   Low Manufacturing Impact
Supplier:   Greiner Bio-One
Microplates, 96-well, non binding
Catalog # 738-0166
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Microplates, 96-well, non binding
Microplates, 96-well, non binding
96-well microplate, V-bottom, non binding, solid bottom, Clear
Price
298,00 €
Log in for availability
1 * 40 ST
Product Details & Documents
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
Specifications
  • Colour:
    Clear
  • Description:
    96-well microplate, V-bottom, non binding, solid bottom
  • Seq:
    02
  • Cat. No.:
    738-0166
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.