Order Entry
Belgium
Orders LinkContactUsLinkComponent
Paraformaldehyde, powder, TECHNICAL

Paraformaldehyde, powder, TECHNICAL

Plastic drum, 25 kg
Catalog # 28794.460
Flammable
Health
Irritant
Supplier:   VWR Chemicals
Paraformaldehyde, powder, TECHNICAL
Catalog # 28794.460
Supplier:  VWR Chemicals
CAS Number:  30525-89-4
Paraformaldehyde, powder, TECHNICAL
Paraformaldehyde, powder, TECHNICAL

Plastic drum, 25 kg

CAS Number:  30525-89-4
Price
447,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Polyoxymethylene
  • CAS Number:30525-89-4
  • Formula:(CH₂O)n
  • MDL Number:MFCD00133991
  • Merck Index:13,07096
  • Boiling Pt:135…136 °C (13 Torr)
  • Melting Pt:120…170 °C
  • Density:0.88 g/cm³ (25 °C)
  • Flash Pt:70 °C (closed cup)
  • Storage Temperature:Ambient
  • UN:2213
  • ADR:4.1,III
Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.