Order Entry
Belgium
Orders LinkContactUsLinkComponent
Laemmli SDS sample buffer, non-reducing (4X)

Laemmli SDS sample buffer, non-reducing (4X)

Laemmli SDS Sample Buffer, non-reducing, 4X, 25 mL
Catalog # J63615.AC
Laemmli SDS sample buffer, non-reducing (4X)
Catalog # J63615.AC
CAS Number:  
Laemmli SDS sample buffer, non-reducing (4X)
Laemmli SDS sample buffer, non-reducing (4X)

Laemmli SDS Sample Buffer, non-reducing, 4X, 25 mL

CAS Number:  
Price
72,50 €
Log in for availability
1 * 25 mL
Product Details & Documents

Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system

Specifications

Specifications

  • Pk:
    25 mL
  • Description:
    Laemmli SDS Sample Buffer, non-reducing, 4X
  • Cat. No.:
    J63615.AC
  • Physical form:
    Liquid
  • Appearance:
    Blue

Specifications