Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)

Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)

TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 4 L
Catalog # J63487.K7
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)
Catalog # J63487.K7
CAS Number:  
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)

TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 4 L

CAS Number:  
Price
102,00 €
Log in for availability
1 * 4 L
Product Details & Documents

Physical Form: Liquid
MDL No.: MFCD00236358

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Description:
    TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer)
  • Cat. No.:
    J63487.K7
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00236358

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.