Phosphate, 0,5M buffer solution, pH 7,5
Phosphate buffer soln, 0,5 M, 7,5, 250 mL
Supplier: Thermo Fisher Scientific
Phosphate, 0,5M buffer solution, pH 7,5
Catalog # J63349.AK
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Specifications
Specifications
- Concentration:0,5 M
- Pk:250 mL
- Description:Phosphate buffer soln
- pH value:7,5
- Cat. No.:J63349.AK
- Physical form:Liquid
- MDL No.:MFCD00003527
- CAS No.:7558-80-7



