Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

IEF Anode buffer (50X)

IEF Anode buffer (50X), 1 L
Catalog # J63303.K2
IEF Anode buffer (50X)
Catalog # J63303.K2
CAS Number:  
IEF Anode buffer (50X)
IEF Anode buffer (50X)

IEF Anode buffer (50X), 1 L

CAS Number:  
Price
50,60 €
Log in for availability
1 * 1 L
Product Details & Documents

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD03846400
Applications: For isoelectric focusing (IEF)

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    IEF Anode buffer (50X)
  • Cat. No.:
    J63303.K2
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD03846400
  • Appearance:
    Colourless

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.