Order Entry
Belgium
Orders LinkContactUsLinkComponent
β-Cyclodextrine

β-Cyclodextrine

Glass bottle for solids, 100 g
Catalog # J63161.22
β-Cyclodextrine
Catalog # J63161.22
CAS Number:  7585-39-9
β-Cyclodextrine
β-Cyclodextrine

Glass bottle for solids, 100 g

CAS Number:  7585-39-9
Price
109,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Use to solubilize non-polar compounds such a fatty acids, lipids and cholesterol. Reported useful for the selective precipitation of enantiomeric, positional or structural isomers

Use to solubilize non-polar compounds such a fatty acids, lipids and cholesterol. Reported useful for the selective precipitation of enantiomeric, positional or structural isomers

Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • Merck Index:14,2718
  • Melting Pt:290 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:231-493-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.