Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

β-Cyclodextrine

Glass bottle for solids, 25 g
Catalog # J63161.14
β-Cyclodextrine
Catalog # J63161.14
CAS Number:  7585-39-9
β-Cyclodextrine
β-Cyclodextrine

Glass bottle for solids, 25 g

CAS Number:  7585-39-9
Price
43,80 €
Log in for availability
1 * 25 g
Product Details & Documents

Use to solubilize non-polar compounds such a fatty acids, lipids and cholesterol. Reported useful for the selective precipitation of enantiomeric, positional or structural isomers

Use to solubilize non-polar compounds such a fatty acids, lipids and cholesterol. Reported useful for the selective precipitation of enantiomeric, positional or structural isomers

Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • Merck Index:14,2718
  • Melting Pt:290 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:231-493-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications