Order Entry
Belgium
Orders LinkContactUsLinkComponent
Neutral red

Neutral red

Glass bottle for solids, 25 g
Catalog # J63054.14
Neutral red
Catalog # J63054.14
CAS Number:  553-24-2
Neutral red
Neutral red

Glass bottle for solids, 25 g

CAS Number:  553-24-2
Price
247,00 €
Log in for availability
1 * 25 g
Product Details & Documents

CAS No.: 553-24-2 Molecular Formula: C15H17ClN4 Formula Weight: 288.78 Physical Form: Powder Appearance: Dark green to black Melting Point: 290° dec. Beilstein Reference: 3918740 Merck Reference: 14,6488 MDL No.: MFCD00012651

Nuclear counterstain useful in pH range 6.8 - 8.0

Synonyms:  N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
  • CAS Number:553-24-2
  • Formula:C₁₅H₁₇ClN₄
  • MDL Number:MFCD00012651
  • Molecular Weight:288.78 g/mol
  • Merck Index:14,6488
  • Melting Pt:290 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:209-035-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.