Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dextran, M.W. ~ 6,000

Dextran, M.W. ~ 6,000

Glass bottle for solids, 50 g
Catalog # J62775.18
Dextran, M.W. ~ 6,000
Catalog # J62775.18
CAS Number:  9004-54-0
Dextran, M.W. ~ 6,000
Dextran, M.W. ~ 6,000

Glass bottle for solids, 50 g

CAS Number:  9004-54-0
Price
134,00 €
Log in for availability
1 * 50 g
Product Details & Documents

CAS No.: 9004-54-0 Molecular Formula: (C6H10O5)n Physical Form: Powder Appearance: White to off-white Merck Reference: 14,2948 MDL No.: MFCD00130935

  • CAS Number:9004-54-0
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00130935
  • Merck Index:14,2948
  • Storage Temperature:Ambient
  • EINECS:232-677-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.