Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
CHAPS lysis buffer

CHAPS lysis buffer

CHAPS lysis buffer, 100 mL
Catalog # J60580.AE
CHAPS lysis buffer
Catalog # J60580.AE
CAS Number:  
CHAPS lysis buffer
CHAPS lysis buffer

CHAPS lysis buffer, 100 mL

CAS Number:  
Price
170,00 €
Connexion Pour la disponibilité
1 * 100 mL
Product Details & Documents

For cell lysis and protein extraction.

Appearance: Clear liquid
MDL No.: MFCD00282866

Contains 50 mM Tris-HCl (pH 7,4), 110 mM NaCl, 5 mM EDTA and 1% CHAPS

Spécifications

Spécifications

  • Pk:
    100 mL
  • Description:
    CHAPS lysis buffer
  • Cat. No.:
    J60580.AE
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00282866
  • Appearance:
    Clear liquid

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.