Order Entry
Belgium
Orders LinkContactUsLinkComponent
CHAPS lysis buffer

CHAPS lysis buffer

CHAPS lysis buffer, 50 mL
Catalog # J60580.AD
CHAPS lysis buffer
Catalog # J60580.AD
CAS Number:  
CHAPS lysis buffer
CHAPS lysis buffer

CHAPS lysis buffer, 50 mL

CAS Number:  
Price
108,00 €
Log in for availability
1 * 50 mL
Product Details & Documents

For cell lysis and protein extraction.

Appearance: Clear liquid
MDL No.: MFCD00282866

Contains 50 mM Tris-HCl (pH 7,4), 110 mM NaCl, 5 mM EDTA and 1% CHAPS

Specifications

Specifications

  • Pk:
    50 mL
  • Description:
    CHAPS lysis buffer
  • Cat. No.:
    J60580.AD
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00282866
  • Appearance:
    Clear liquid

Specifications