Laemmli SDS sample buffer, reducing (6X)
Laemmli SDS Sample Buffer, reducing, 6X, 25 mL
Supplier: Thermo Fisher Scientific
Laemmli SDS sample buffer, reducing (6X)
Catalog # J61337.AC
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system
Documents
Specifications
Specifications
- Pk:25 mL
- Description:Laemmli SDS Sample Buffer, reducing, 6X
- Cat. No.:J61337.AC
- Physical form:Liquid
- Appearance:Blue



