Order Entry
Belgium
Orders LinkContactUsLinkComponent
EPPS, 0,2 M buffer solutions

EPPS, 0,2 M buffer solutions

EPPS buffer solution, 0,2 mol/L, 8,0, Glass bottle, 250 mL
Catalog # J61296.AK
EPPS, 0,2 M buffer solutions
Catalog # J61296.AK
CAS Number:  
EPPS, 0,2 M buffer solutions
EPPS, 0,2 M buffer solutions

EPPS buffer solution, 0,2 mol/L, 8,0, Glass bottle, 250 mL

CAS Number:  
Price
148,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

CAS No.: 16052-06-5
Physical form: liquid
MDL No.: MFCD00006160

Specifications

Specifications

  • Concentration:
    0,2 mol/L
  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Description:
    EPPS buffer solution
  • pH value:
    8,0
  • Cat. No.:
    J61296.AK
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00006160
  • CAS No.:
    16052-06-5

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.