Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

EPPS, 0,2 M buffer solutions

EPPS buffer solution, 0,2 mol/L, 8,0, Glass bottle, 100 mL
Catalog # J61296.AE
EPPS, 0,2 M buffer solutions
Catalog # J61296.AE
CAS Number:  
EPPS, 0,2 M buffer solutions
EPPS, 0,2 M buffer solutions

EPPS buffer solution, 0,2 mol/L, 8,0, Glass bottle, 100 mL

CAS Number:  
Price
103,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

CAS No.: 16052-06-5
Physical form: liquid
MDL No.: MFCD00006160

Specifications

Specifications

  • Concentration:
    0,2 mol/L
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    EPPS buffer solution
  • pH value:
    8,0
  • Cat. No.:
    J61296.AE
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00006160
  • CAS No.:
    16052-06-5

Specifications