HEPES, 1,0 M buffer solutions
HEPES buffer solution, 1 mol/L, 7,0, Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 7365-45-9
MDL No.: MFCD00006158
Specifications
Specifications
- Concentration:1 mol/L
- Pack type:Glass bottle
- Pk:500 mL
- Description:HEPES buffer solution
- pH value:7,0
- Cat. No.:J62688.AP
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00006158
- Appearance:Colourless
- CAS No.:7365-45-9



