Tricine-SDS sample buffer (2X), non-reducing
Tricine-SDS Sample Buffer (2X), non-reducing, 50 mL
Supplier: Thermo Fisher Scientific
Tricine-SDS sample buffer (2X), non-reducing
Catalog # J62677.AD
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid
Appearance: Blue
Specifications
Specifications
- Pk:50 mL
- Description:Tricine-SDS Sample Buffer (2X), non-reducing
- Cat. No.:J62677.AD
- Physical form:Liquid
- Appearance:Blue
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



