Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Tricine-SDS sample buffer (2X), non-reducing

Tricine-SDS Sample Buffer (2X), non-reducing, 25 mL
Catalog # J62677.AC
Tricine-SDS sample buffer (2X), non-reducing
Catalog # J62677.AC
CAS Number:  
Tricine-SDS sample buffer (2X), non-reducing
Tricine-SDS sample buffer (2X), non-reducing

Tricine-SDS Sample Buffer (2X), non-reducing, 25 mL

CAS Number:  
Price
49,80 €
Log in for availability
1 * 25 mL
Product Details & Documents

Physical Form: Liquid
Appearance: Blue

Specifications

Specifications

  • Pk:
    25 mL
  • Description:
    Tricine-SDS Sample Buffer (2X), non-reducing
  • Cat. No.:
    J62677.AC
  • Physical form:
    Liquid
  • Appearance:
    Blue

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.