ADA, 0,2 M Buffer Solutions
ADA buffer solution, 0,2 M, 6,5, Glass bottle, 250 mL
N-(2-Acetimido)iminodiacetic acid monosodium salt (ADA) buffer solutions PH values 6,0 to 7,5.
CAS No.: 7415-22-7
Appearance: Colourless
MDL No.: MFCD00065483
: Storage Temperature: 2 to 8°C
- Concentration:0,2 M
- Pack type:Glass bottle
- Pk:250 mL
- Description:ADA buffer solution
- pH value:6,5
- Cat. No.:J61259.AK
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00065483
- Appearance:Colourless
- CAS No.:7415-22-7



