ADA, 0,2 M Buffer Solutions
ADA buffer solution, 0,2 M, 6,5, Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
N-(2-Acetimido)iminodiacetic acid monosodium salt (ADA) buffer solutions PH values 6,0 to 7,5.
CAS No.: 7415-22-7
Appearance: Colourless
MDL No.: MFCD00065483
: Storage Temperature: 2 to 8°C
Specifications
Specifications
- Concentration:0,2 M
- Pack type:Glass bottle
- Pk:100 mL
- Description:ADA buffer solution
- pH value:6,5
- Cat. No.:J61259.AE
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00065483
- Appearance:Colourless
- CAS No.:7415-22-7
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



