Order Entry
Belgium
Orders LinkContactUsLinkComponent
Paraformaldehyde fixative solution

Paraformaldehyde fixative solution

500 mL
Catalog # J61984.AP
Health
Irritant
Paraformaldehyde fixative solution
Catalog # J61984.AP
CAS Number:  30525-89-4
Paraformaldehyde fixative solution
Paraformaldehyde fixative solution

500 mL

CAS Number:  30525-89-4
Price
181,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Paraformaldehyde fixative solution finds application in proteins, peptides and the enzymes of low molecular weight.

Storage Temperature: 2°C to 8°C Physical Form: Liquid MDL No.: MFCD00133991

Synonyms:  Polyoxymethylene
  • CAS Number:30525-89-4
  • Formula:(CH₂O)n
  • MDL Number:MFCD00133991
Specifications

Specifications

  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.