Paraformaldehyde fixative solution
250 mL
Supplier: Thermo Fisher Scientific
Paraformaldehyde fixative solution
Catalog # J61984.AK
Supplier: Thermo Fisher Scientific
CAS Number: 30525-89-4
Jump to:
Product Details & Documents
Paraformaldehyde fixative solution finds application in proteins, peptides and the enzymes of low molecular weight.
Storage Temperature: 2°C to 8°C Physical Form: Liquid MDL No.: MFCD00133991
Synonyms: Polyoxymethylene
- CAS Number:30525-89-4
- Formula:(CH₂O)n
- MDL Number:MFCD00133991
Documents
Specifications
Specifications
- Pk:250 mL



