Order Entry
Belgium
Orders LinkContactUsLinkComponent
GTE buffer

GTE buffer

100 mL
Catalog # J62597.AE
GTE buffer
Catalog # J62597.AE
CAS Number:  
GTE buffer
GTE buffer

100 mL

CAS Number:  
Price
41,90 €
Log in for availability
1 * 100 mL
Product Details & Documents

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Applications: For miniprep and DNA purification

Specifications

Specifications

  • Pk:
    100 mL
  • Cat. No.:
    J62597.AE
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid

Specifications