Order Entry
Belgium
Orders LinkContactUsLinkComponent
Nitrocellulose stripping buffer (10X)

Nitrocellulose stripping buffer (10X)

Nitrocellulose Stripping Buffer, 10X, 500 mL
Catalog # J62541.AP
Nitrocellulose stripping buffer (10X)
Catalog # J62541.AP
CAS Number:  
Nitrocellulose stripping buffer (10X)
Nitrocellulose stripping buffer (10X)

Nitrocellulose Stripping Buffer, 10X, 500 mL

CAS Number:  
Price
98,40 €
Log in for availability
1 * 500 mL
Product Details & Documents

Physical Form: Liquid

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Nitrocellulose Stripping Buffer, 10X
  • Cat. No.:
    J62541.AP
  • Physical form:
    Liquid

Specifications