Order Entry
Belgium
Orders LinkContactUsLinkComponent
TBE sample buffer (6X)

TBE sample buffer (6X)

TBE Sample Buffer, 6X, 25 mL
Catalog # J60357.AC
TBE sample buffer (6X)
Catalog # J60357.AC
CAS Number:  
TBE sample buffer (6X)
TBE sample buffer (6X)

TBE Sample Buffer, 6X, 25 mL

CAS Number:  
Price
55,00 €
Log in for availability
1 * 25 mL
Product Details & Documents

Physical Form: Liquid
Appearance: Blue

Specifications

Specifications

  • Pk:
    25 mL
  • Description:
    TBE Sample Buffer, 6X
  • Cat. No.:
    J60357.AC
  • Physical form:
    Liquid
  • Appearance:
    Blue

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.