Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bacitracin

Bacitracin

Glass bottle for solids, 5 g
Catalog # J62432.06
Bacitracin
Catalog # J62432.06
CAS Number:  1405-87-4
Bacitracin
Bacitracin

Glass bottle for solids, 5 g

CAS Number:  1405-87-4
Price
80,60 €
Log in for availability
1 * 5 g
Product Details & Documents

Inhibits cell wall synthesis in Gram-positive bacteria.

  • White to off-white powder

Soluble in water, methanol or ethanol

  • CAS Number:1405-87-4
  • Formula:C₆₆H₁₀₃N₁₇O₁₆S₁
  • MDL Number:MFCD00062640
  • Molecular Weight:1421.60 g/mol
  • Merck Index:14,934
  • Melting Pt:222 °C
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications