Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Phosphate, 0,2 M buffer solution, pH 7,2

Phosphate buffer soln, 0,2 M, 7,2, 1 L
Catalog # J62899.K2
Phosphate, 0,2 M buffer solution, pH 7,2
Catalog # J62899.K2
CAS Number:  
Phosphate, 0,2 M buffer solution, pH 7,2
Phosphate, 0,2 M buffer solution, pH 7,2

Phosphate buffer soln, 0,2 M, 7,2, 1 L

CAS Number:  
Price
108,00 €
Log in for availability
1 * 1 L
Product Details & Documents

CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Specifications

Specifications

  • Concentration:
    0,2 M
  • Pk:
    1 L
  • Description:
    Phosphate buffer soln
  • pH value:
    7,2
  • Cat. No.:
    J62899.K2
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00003527
  • CAS No.:
    7558-80-7

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.