Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

TBE urea sample buffer (2X)

TBE Urea Sample Buffer, 2X, 10 mL
Catalog # J60186.AB
TBE urea sample buffer (2X)
Catalog # J60186.AB
CAS Number:  
TBE urea sample buffer (2X)
TBE urea sample buffer (2X)

TBE Urea Sample Buffer, 2X, 10 mL

CAS Number:  
Price
79,80 €
Log in for availability
1 * 10 mL
Product Details & Documents

Physical Form: Liquid
Appearance: Blue

Specifications

Specifications

  • Pk:
    10 mL
  • Description:
    TBE Urea Sample Buffer, 2X
  • Cat. No.:
    J60186.AB
  • Physical form:
    Liquid
  • Appearance:
    Blue

Specifications