Order Entry
Belgium
Orders LinkContactUsLinkComponent
Separating or resolving buffer (4X)

Separating or resolving buffer (4X)

Separating or Resolving Buffer, 4X, 1 L
Catalog # J60181.K2
Separating or resolving buffer (4X)
Catalog # J60181.K2
CAS Number:  
Separating or resolving buffer (4X)
Separating or resolving buffer (4X)

Separating or Resolving Buffer, 4X, 1 L

CAS Number:  
Price
114,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Physical Form: Liquid
Appearance: Clear

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Separating or Resolving Buffer, 4X
  • Cat. No.:
    J60181.K2
  • Physical form:
    Liquid
  • Appearance:
    Clear

Specifications